• USA Home
  • 529524 - Potassium hexafluorogermanate

EMAIL THIS PAGE TO A FRIEND
529524 Aldrich

Potassium hexafluorogermanate

Purchase

Properties

form   powder
bp   835 °C(lit.)
mp   730 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?