EMAIL THIS PAGE TO A FRIEND
577898 Supelco

Methanol solution

contains 0.1 % (v/v) trifluoroacetic acid

  • CAS Number 67-56-1

  • Empirical Formula (Hill Notation) CH4O

  • Molecular Weight 32.04

  •  Beilstein/REAXYS Number 3662861

  •  EC Number 200-659-6

  •  MDL number MFCD00004595

  •  PubChem Substance ID 24880898

Purchase

Properties

Related Categories Amber Glass Bottles, HPLC Blends, HPLC Grade Solvents, HPLC Solvents, Pre-Blended Mobile Phase Solvents,
Quality Level   100
vapor density   1.11 (vs air)
vapor pressure   410 mmHg
contains   0.1 % (v/v) trifluoroacetic acid
expl. lim.   5.5-44 % (lit.)
bp   65.4 °C (lit.)
mp   -98 °C (lit.)
SMILES string   CO
InChI   1S/CH4O/c1-2/h2H,1H3
InChI key   OKKJLVBELUTLKV-UHFFFAOYSA-N

Description

Packaging

4×4 L packaged in glass bottles; 18 L packaged in VerSA-Flow® containers

Legal Information

Pure-Pac is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
WGK 1
Flash Point(F) 
51.8 °F - closed cup - Not applicable
Flash Point(C) 
11 °C - closed cup - Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Dispensette™ S
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?