EMAIL THIS PAGE TO A FRIEND
587036 Sigma-Aldrich

Sodium formate-12C

99.9 atom % 12C

Synonym: Formic acid-12C sodium salt

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, Q-S, Stable Isotopes More...
description   13C depleted
isotopic purity   99.9 atom % 12C
quality   13C depleted
mp   259-262 °C (lit.)
mass shift   depleted
SMILES string   [Na+].[O-][12CH]=O
InChI   1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1/i1+0;
InChI key   HLBBKKJFGFRGMU-XUGDXVOUSA-M

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?