EMAIL THIS PAGE TO A FRIEND
591165 Supelco

Water solution

contains 0.10 % (v/v) acetic acid

Purchase

Description

Packaging

200 L packaged in closed-head composite drums; 4×4 L packaged in glass bottles; 18 L packaged in VerSA-Flow® containers

Legal Information

Pure-Pac is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Dispensette™ S
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?