• USA Home
  • 597503 - Acetonitrile solution

EMAIL THIS PAGE TO A FRIEND
597503 Sigma-Aldrich

Acetonitrile solution

contains 0.035 % (v/v) acetic acid

Purchase

Properties

Quality Level   100
contains   0.035 % (v/v) acetic acid
density   0.782 g/mL at 25 °C
SMILES string   CC#N
InChI   1S/C2H3N/c1-2-3/h1H3
InChI key   WEVYAHXRMPXWCK-UHFFFAOYSA-N

Description

Packaging

4×4 L packaged in glass bottles; 18 L packaged in VerSA-Flow® containers

Legal Information

Pure-Pac is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
WGK 2
Flash Point(F) 
42.0 °F - Not applicable
Flash Point(C) 
5.55 °C - Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?