• USA Home
  • 603740 - Allyl chloride-1-13C

EMAIL THIS PAGE TO A FRIEND
603740 Sigma-Aldrich

Allyl chloride-1-13C

99 atom % 13C

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories A, Alphabetical Listings, Stable Isotopes More...
description   random-1,3-13C1
isotopic purity   99 atom % 13C
bp   44-46 °C (lit.)
mp   -130 °C (lit.)
density   0.951 g/mL at 25 °C
mass shift   M+1
SMILES string   Cl[13CH2]C=C
InChI   1S/C3H5Cl/c1-2-3-4/h2H,1,3H2/i3+1
InChI key   OSDWBNJEKMUWAV-LBPDFUHNSA-N

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1100 - class 3 - PG 1 - Allyl chloride
WGK Germany 
3
Flash Point(F) 
-25.6 °F
Flash Point(C) 
-32 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?