EMAIL THIS PAGE TO A FRIEND
608734 Sigma-Aldrich

Urea-14N2

99.99 atom % 14N, 15N-depleted

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, Stable Isotopes, U-Z More...
isotopic purity   99.99 atom % 14N
mp   132-135 °C (lit.)
mass shift   depleted
SMILES string   [14NH2]C([14NH2])=O
InChI   1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i2+0,3+0
InChI key   XSQUKJJJFZCRTK-VTTDIVQUSA-N

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YR6250000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?