EMAIL THIS PAGE TO A FRIEND
640271 Supelco

Water solution

HPLC grade, contains 0.035 % (v/v) trifluoroacetic acid

Purchase

Properties

grade   HPLC grade
contains   0.035 % (v/v) trifluoroacetic acid
SMILES string   O
InChI   1S/H2O/h1H2
InChI key   XLYOFNOQVPJJNP-UHFFFAOYSA-N

Description

Packaging

4×4 L packaged in glass bottles; 18 L packaged in VerSA-Flow® containers

Legal Information

Pure-Pac is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?