• USA Home
  • 643769 - (3,5,5-Trimethylcyclohex-2-enylidene)malononitrile

EMAIL THIS PAGE TO A FRIEND
643769 Sigma-Aldrich

(3,5,5-Trimethylcyclohex-2-enylidene)malononitrile

  • CAS Number 23051-44-7

  • Empirical Formula (Hill Notation) C12H14N2

  • Molecular Weight 186.25

  •  MDL number MFCD00228058

  •  PubChem Substance ID 24883294

  •  NACRES NA.23

Purchase

Properties

Related Categories Materials Science, Monomers, Polymer Science, Vinyl Halides, Amines, Amides, and Other Vinyl Monomers
mp   73-77 °C (lit.)
SMILES string   CC1=CC(\CC(C)(C)C1)=C(\C#N)C#N
InChI   1S/C12H14N2/c1-9-4-10(11(7-13)8-14)6-12(2,3)5-9/h4H,5-6H2,1-3H3
InChI key   QTUUBOZSGQFLRZ-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Features and Benefits

Intermediate in dye preparations.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?