• USA Home
  • 652431 - Neodymium(II) iodide

EMAIL THIS PAGE TO A FRIEND
652431 Sigma-Aldrich

Neodymium(II) iodide

anhydrous, powder, ≥99.9% trace metals basis

Synonym: Neodymium diiodide

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Crystal Grade Inorganics, Essential Chemicals, Materials Science,
grade   anhydrous
InChI Key   CTHFQDYUBRIODB-UHFFFAOYSA-L
assay   ≥99.9% trace metals basis
form   powder
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles

Articles

The Rare Earth Crisis—The Supply/Demand Situation for 2010–2015

K.A. Gschneidner, Jr. Ames Laboratory, U.S. Department of Energy and Department of Materials Science and Engineering Iowa State University, Ames, Iowa 50011-3020, USA Email: cagey@ameslab.gov
Keywords: Absorption, Automotive, Capillary electrophoresis, Catalysis, Ceramics, Coloring agents, Detection methods, Environmental, Industries, Magnetic resonance imaging, Materials Science, Oxidations, Poisons, Reductions, Scintillation, Semiconductor, Separation, Substitutions

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?