• USA Home
  • 679216 - Ethyltrichlorosilane

EMAIL THIS PAGE TO A FRIEND
679216 Aldrich

Ethyltrichlorosilane

deposition grade, ≥98% (GC), ≥99.99% (as metals)

Synonym: Trichloro(ethyl)silane

  • CAS Number 115-21-9

  • Empirical Formula (Hill Notation) C2H5Cl3Si

  • Molecular Weight 163.51

  •  Beilstein Registry Number 1361384

  •  EC Number 204-072-6

  •  MDL number MFCD00000485

  •  PubChem Substance ID 24885476

Purchase

Properties

Related Categories Chemical Synthesis, Materials Science, Micro/NanoElectronics, Protecting and Derivatizing Reagents, Protection and Derivatization,
grade   deposition grade
InChI Key   ZOYFEXPFPVDYIS-UHFFFAOYSA-N
assay   ≥98% (GC)
  ≥99.99% (as metals)
form   liquid
refractive index   n20/D 1.425(lit.)
bp   99 °C(lit.)
mp   −106 °C(lit.)
density   1.238 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Corrosive to the respiratory tract., Reacts violently with water.
RIDADR 
UN 1196 8(3) / PGII
WGK Germany 
1
RTECS 
VV4200000
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles

Articles

Deposition Grade Silanes for Sol-Gel Processes

Sol-gel processing is beneficial in the formation of ceramic and glass films for a number of reasons.1-4 It is a simple process that does not require exotic materials, catalysts or expensive depositi...
Keywords: Chromatography, Condensations, Deposition, Electronics, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Nuclear magnetic resonance spectroscopy, Sol-gel

Sol-Gel Science for Ceramic Materials

Silicon-based research has drawn much attention in recent years with targeted organizations such as the International Sol-Gel Society hosting workshops,1 and the Sol-Gel Gateway2 hosting a compendium...
Dr. Sandra Kay Young
Material Matters 2006, 1.3, 8.
Keywords: Ceramics, Condensations, Deposition, Evaporation, Hydrosilylations, Infrared spectroscopy, Ligands, Nuclear magnetic resonance spectroscopy, Polymerization reactions, Sol-gel

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.1 Since then, the number of hybrid organicinorganic combinations has increased rapidly.2 Hybrid mater...
Lisa C. Klein and Andrei Jitianu
Material Matters, Volume 7, Number 2, 8–14
Keywords: Absorption, Adhesion, Calorimetry, Combustion, Condensations, Deposition, Exothermic, Filtration, Materials Science, Melting, Oxidations, Polymerization reactions, Purification, Semiconductor, Sol-gel, Solar cells, Solvents, Spin coating, Substitutions, Thermal analysis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?