• USA Home
  • 693154 - (S)-[(RuCl(DM-BINAP))2(μ-Cl)3][NH2Me2]

EMAIL THIS PAGE TO A FRIEND
693154 Aldrich

(S)-[(RuCl(DM-BINAP))2(μ-Cl)3][NH2Me2]

Synonym: Dimethylammonium dichlorotri(μ-chloro)bis[(S)-(−)-2,2′-bis[di-(3,5-xylyl)phosphino]-1,1′-binaphthyl]diruthenate(II)

Purchase

Properties

Related Categories Asymmetric Synthesis, BINAPs, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Privileged Ligands and Complexes More...
InChI Key   LGSCCUXQQTYWAH-UHFFFAOYSA-M
storage temp.   2-8°C

Description

Legal Information

Sold in collaboration with Takasago for research purposes only.

Application

Takasago Ligands and Complexes for Asymmetric Reactions

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles

Articles

Takasago Ligands and Complexes for Asymmetric Reactions

The quest to find chiral catalysts capable of achieving high enantiomeric excess for a variety of asymmetric transformations has been an ongoing endeavor for the past 40 years. One of the most releva...
Keywords: Addition reactions, Arylations, Asymmetric synthesis, Catalysis, Hydrogenations, Ligands, transformation

Related Content

Takasago Ligands - Advantages and Applications

Introduction The quest to find chiral catalysts capable of achieving high enantiomeric excess for a variety of asymmetric transformations has been an ongoing endeavor for the past 40 years. One of th...
Keywords: Addition reactions, Applications, Arylations, Asymmetric synthesis, Building blocks, Catalysis, Epoxidations, Hydrogenations, Hydrosilylations, Ligands, Sharpless Epoxidation, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?