• USA Home
  • 709786 - 4-Methylbenzenethioamide

EMAIL THIS PAGE TO A FRIEND
709786 Sigma-Aldrich

4-Methylbenzenethioamide

96%

  • CAS Number 2362-62-1

  • Empirical Formula (Hill Notation) C8H9NS

  • Molecular Weight 151.23

  •  MDL number MFCD00173750

  •  PubChem Substance ID 329762896

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocarbonyl Compounds More...
assay   96%
mp   170-174 °C
storage temp.   room temp
SMILES string   Cc1ccc(cc1)C(N)=S
InChI   1S/C8H9NS/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3,(H2,9,10)
InChI key   QXYZSNGZMDVLKN-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?