• USA Home
  • 720232 - 4-Amino-2-fluoropyridine

EMAIL THIS PAGE TO A FRIEND
720232 Sigma-Aldrich

4-Amino-2-fluoropyridine

96%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Fluorinated Building Blocks, Heterocyclic Fluorinated Building Blocks, Other Fluorinated Heterocycles,
assay   96%
mp   83-88 °C
storage temp.   2-8°C
SMILES string   Nc1ccnc(F)c1
InChI   1S/C5H5FN2/c6-5-3-4(7)1-2-8-5/h1-3H,(H2,7,8)
InChI key   JSAKBYXSHKYFQU-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?