• USA Home
  • 752290 - Molybdenum disilicide

EMAIL THIS PAGE TO A FRIEND
752290 Aldrich

Molybdenum disilicide Green Alternative

α-phase, powder, ≥99.8% trace metals basis

Synonym: Molybdenum silicide

Purchase

Properties

Related Categories 3D Printing Materials for Research and Development, Alternative Energy, Ceramics for 3D Printing, Chemical Synthesis, Enabling Products,
InChI Key   OVVBPJHTRZEJHC-UHFFFAOYSA-N
assay   ≥99.8% trace metals basis
form   powder
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
avg. part. size   7.5 μm
mp   1870-2030 °C (lit.)
density   6.31 g/mL at 25 °C(lit.)

Description

Packaging

25 g in glass bottle

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

3D Printing of Carbon Fiber-Reinforced Composites

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries, most typically using a layer-by-layer fabrication process. 3D pri...
Zhenyu Bo* (Ph.D Candidate at Northwestern University) and Jia Choi*, PhD, Product Manager


*Materials Science Product Management Team, MilliporeSigma, Milwaukee, WI.
Keywords: Deposition, Nanomaterials, Nanotubes

Silicides: Promising Thermoelectric Materials - Material Matters Volume 6 Number 4

In recent years, the price of tellurium, a key component in the best-performing thermoelectric materials, has increased significantly, leading to the question, "Is it economically viable to produce t...
Mikhail I. Fedorov, Vladimir K. Zaitsev
Material Matters Volume 6 Number 4
Keywords: Applications, Gene expression, Melting, Methods, Nucleic acid annealing, Reductions, Semiconductor, Type

Silicides: Thermoelectric Materials

Mikhail I. Fedorov,* Vladimir K. Zaitsev Ioffe Physical-technical Institute, St. Petersburg, Russia *Email: m.fedorov@mail.ioffe.ru
Keywords: Gene expression, Melting, Nucleic acid annealing, Reductions, Semiconductor

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?