• USA Home
  • 762903 - N,N′-Di(2-naphthyl-N,N′-diphenyl)-1,1′-biphenyl-4,4′-diamine

EMAIL THIS PAGE TO A FRIEND
762903 Sigma-Aldrich

N,N′-Di(2-naphthyl-N,N′-diphenyl)-1,1′-biphenyl-4,4′-diamine

99% (HPLC)

Synonym: N,N′-Bis(naphthalen-2-yl)-N,N′-bis(phenyl)benzidine, beta-NPB

  • CAS Number 139255-17-7

  • Empirical Formula (Hill Notation) C44H32N2

  • Molecular Weight 588.74

  •  MDL number MFCD03093247

  •  PubChem Substance ID 329767010

  •  NACRES NA.23

Purchase

Properties

Related Categories Hole Transport (HT) & Hole Injection Layer (HIL) Materials, Hole Transport Materials, Materials Science, OLED and PLED Materials, Organic and Printed Electronics More...
Quality Level   100
assay   99% (HPLC)
form   powder
mp   168-175 °C
SMILES string   c1ccc(cc1)N(c2ccc(cc2)-c3ccc(cc3)N(c4ccccc4)c5ccc6ccccc6c5)c7ccc8ccccc8c7
InChI   1S/C44H32N2/c1-3-15-39(16-4-1)45(43-29-23-33-11-7-9-13-37(33)31-43)41-25-19-35(20-26-41)36-21-27-42(28-22-36)46(40-17-5-2-6-18-40)44-30-24-34-12-8-10-14-38(34)32-44/h1-32H
InChI key   BLFVVZKSHYCRDR-UHFFFAOYSA-N

Description

Application

Used in organic electronic light emitting devices.

This material is used as a hole transport layer in a variety of OLED devices

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Aggregation-induced Emission Luminogens for Non-doped Organic Light-emitting Diodes

Han Nie,1 Zujin Zhao1a and Ben Zhong Tang1,2,3b 1State Key Laboratory of Luminescent Materials and Devices, South China University of Technology, Guangzhou 510640, China 2Department of Chemistry, The...
Han Nie, Zujin Zhao, Ben Zhong Tang
Material Matters, 2016, 11.1, 29
Keywords: Bacterial conjugations, Building blocks, Deposition, Electronics, Indicators, Organic electronics, Semiconductor, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?