• USA Home
  • 767182 - Polyester bis-MPA dendron, 32 hydroxyl, 1 azide

EMAIL THIS PAGE TO A FRIEND
767182 Sigma-Aldrich

Polyester bis-MPA dendron, 32 hydroxyl, 1 azide

generation 5, 95%

Synonym: bisMPA dendrimer, bisMPA dendron

  •  NACRES NA.23

Purchase

Properties

Related Categories Dendrimers, Dendrons, Materials Science, Nanomaterials
Quality Level   100
description   hydroxyl surface groups
assay   95%
mol wt   generation 5
feature   focal point azide
no. Surface Groups   32
storage temp.   −20°C

Description

Application

Azide dendrons can be coupled to acetylene functionalized molecules, including acetylene dendrons to form asymmetric dendrimers.

calculated mol. wt. 3742.8

Packaging

250 mg in glass insert

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Biocompatible Dendritic Building Blocks for Advanced Biomedical Research

Dendrimers and dendrons are symmetrically branched polymer structures that possess a well-defined spatial distribution of functional groups. These dendritic materials have attracted considerable inte...
1Dr. Michael Malkoch and 2Dr. Andreas M. Nyström
Material Matters, 2012 v7, n3
Keywords: Applications, Asymmetric synthesis, Bacterial conjugations, Building blocks, Cancer, Catalysis, Chemical reactions, Click chemistry, Diagnostic, Diagnostic Imaging, Materials Science, Nanomaterials, Solvents, Type

Related Content

Dendritic Polyester Scaffolds for Drug Delivery Applications

Dendrimers and dendrons are highly branched polymer structures with a precise distribution of functional groups. In contrast to their linear analogues, they are monodisperse in nature and present no ...
Sandra García-Gallego
Marie Sklowdoska-Curie Researcher at KTH Royal Institute of Technology, and Michael Malkoch, Associate Professor at KTH Royal Institute of Technology and Chief Executive Officer at Polymer Factory Sweden AB
Keywords: Antibiotics, Bacterial conjugations, Building blocks, Catalysis, Chemical reactions, Chromatography, Click chemistry, Column chromatography, Esterifications, Green chemistry, Nuclear magnetic resonance spectroscopy, Purification, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?