EMAIL THIS PAGE TO A FRIEND
776696 Aldrich

Germanium telluride Green Alternative

99.999% trace metals basis

Synonym: Germanium monotelluride, Germanium(II) telluride, Germanous telluride

Purchase

Properties

Related Categories Alternative Energy, Chalcogenides, Chemical Synthesis, Enabling Products, Greener Alternative Products,
InChI Key   JBQYATWDVHIOAR-UHFFFAOYSA-N
assay   99.999% trace metals basis
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
mp   725 °C(lit.)
density   6.14 g/mL at 25 °C(lit.)

Description

Application

GeTe is a promising material for thermoelectric applications. Nanostructuring; non-stoichiometric defects; doping and alloying with other materials further enhances its thermoelectric performance.
GeTe is also an important semiconductor material prototypical phase change material of high interest for applications in optical and electronic non-volatile memories.

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?