EMAIL THIS PAGE TO A FRIEND
89502 Aldrich

Titanium(II) oxide

coating quality, granular, 99.5%

Synonym: Titanium monoxide

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science, Oxides,
grade   coating quality
InChI Key   OGIDPMRJRNCKJF-UHFFFAOYSA-N
assay   99.5%
form   granular
mfr. no.   Umicore Materials AG
particle size   0.7-1.5 mm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?