• USA Home
  • CDS001167 - 4-N-Butylbenzenesulfonamide

EMAIL THIS PAGE TO A FRIEND
CDS001167 Sigma-Aldrich

4-N-Butylbenzenesulfonamide

AldrichCPR

  • CAS Number 1135-00-8

  • Empirical Formula (Hill Notation) C10H15NO2S

  • Molecular Weight 213.30

  •  MDL number MFCD00465202

  •  PubChem Substance ID 329778391

Purchase

Properties

SMILES string   CCCCc1ccc(cc1)S(N)(=O)=O
InChI   1S/C10H15NO2S/c1-2-3-4-9-5-7-10(8-6-9)14(11,12)13/h5-8H,2-4H2,1H3,(H2,11,12,13)
InChI key   IWEMSJAOIKQBBZ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?