• USA Home
  • CHR101820 - Chemrus® disposable filter funnel
EMAIL THIS PAGE TO A FRIEND
CHR101820
Chemrus® disposable filter funnel
conical, pore size 10 μm, capacity 20 mL
  •  NACRES NB.43
Purchase
Properties
material   polyethylene frit
  polypropylene body
mfr. no.   Chemrus, CR-1018-20
capacity   20 mL
pore size   10 μm
Description
General description
Convenient and inexpensive, these disposable filter funnels have polypropylene bodies with tapered stems that fit 7/10 joints without seals. The funnels are available in three styles, Buchner and Hirsch, and conical, choice of polyethylene frit, perforated plate, acid washed Celite or silica gel. Maximum use temperature is 110 °C. Funnels have an 80 mm long stem so the glass vacuum adapters are not contaminated during the filtration process.
Order vacuum adapters, filter paper, vials and 20-400 to 24-400 vial connectors separately.
Disposable filter funnel s with a polypropylene body with a 10 micron polyethylene fritted disc.
Legal Information
Chemrus is a registered trademark of Liu, Xioagao & Cai, Jianjian
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?