EMAIL THIS PAGE TO A FRIEND
F4043 Sigma-Aldrich

Fisetin

≥98%

Synonym: 3,3′,4′,7-Tetra­hydroxy­flavone, 5-Deoxyquercetin, Natural Brown 1

  • CAS Number 345909-34-4

  • Empirical Formula (Hill Notation) C15H10O6 · xH2O

  • Molecular Weight 286.24 (anhydrous basis)

  •  Colour Index Number 75620

  •  Beilstein/REAXYS Number 292829

  •  EC Number 208-434-4

  •  MDL number MFCD04220857

  •  PubChem Substance ID 57654179

  •  NACRES NA.22

Purchase

Properties

Related Categories Acacia, Benzopyrans, Building Blocks, Cancer Research, Cell Biology,
Quality Level   100
assay   ≥98%
mp   >330 °C (lit.)
storage temp.   −20°C
SMILES string   O.Oc1ccc2C(=O)C(O)=C(Oc2c1)c3ccc(O)c(O)c3
InChI   1S/C15H10O6.H2O/c16-8-2-3-9-12(6-8)21-15(14(20)13(9)19)7-1-4-10(17)11(18)5-7;/h1-6,16-18,20H;1H2
InChI key   GYHFUROKCOMWNQ-UHFFFAOYSA-N
Gene Information   human ... CDC2(983), CDK5(1020), CDK6(1021), CYP1A2(1544), GSK3A(2931)
mouse ... Hexa(15211)
rat ... Il4(287287), Tnf(24835)

Description

General description

Fisetin is a dietary flavonoid found in many fruits and vegetables. It is shown to exhibit anti-cancer, anti-inflammatory, antioxidant activities and also enhance the memory.

Application

Fisetin can be used:
• To prepare its iron metal complexes possessing promising antioxidant activity.
• In the study of its interaction with cyclodextrins (α and β) using analytical tools.

Fisetin is a naturally occurring therapeutically active flavonol which has been used in the synthesis of pharmaceutically active anti-inflammatory, anticonvulsant and antiproliferative agents.

Packaging

100, 500 mg in poly bottle

Other Notes

Brown pigment found in many species of woody plants

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LK9250000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Articles

Dietary Antioxidants

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.1 During the oxi...
Biofiles Volume 2 Article 2
Keywords: Adhesion, Anti-inflammatory agents, Applications, Cancer, Cardiovascular, Catalysis, Diabetes, Diseases, Gene expression, Metabolism, Neurodegenerative Diseases, Peroxidations, Phosphorylations, Prebiotics, Probiotics, Reductions, Transcription, Transduction, Vitamins

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?