EMAIL THIS PAGE TO A FRIEND
GF47689879 Sigma-Aldrich

Molybdenum

foil, light tested, 200x200mm, thickness 0.025mm, annealed, 99.9%

Synonym: MO000220, Molybdenum

Purchase

Properties

assay   99.9%
form   foil
mfr. no.   Goodfellow, 476-898-79
resistivity   5.0 μΩ-cm, 20°C
size × thickness   200 x 200 mm × 0.025 mm
bp   4612 °C (lit.)
mp   2617 °C (lit.)
density   10.3 g/mL at 25 °C (lit.)
SMILES string   [Mo]
InChI   1S/Mo
InChI key   ZOKXTWBITQBERF-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?