EMAIL THIS PAGE TO A FRIEND
GF64590239 Aldrich

Iridium

powder, 850 max. part. size (micron), weight 2 g, high purity 99.995%

Synonym: IR006010

Purchase

Properties

assay   99.995%
form   powder
mfr. no.   Goodfellow, 645-902-39
resistivity   4.71 μΩ-cm
weight   2 g
max. part. size   850 (micron)
bp   4130 °C (lit.)
mp   2450 °C (lit.)
density   22.65 g/cm3 (lit.)
SMILES string   [Ir]
InChI   1S/Ir
InChI key   GKOZUEZYRPOHIO-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?