EMAIL THIS PAGE TO A FRIEND
T34908 Sigma-Aldrich

D-Thyroxine

99%

Synonym: 3,3′,5,5′-Tetraiodo-D-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-D-alanine

  • CAS Number 51-49-0

  • Empirical Formula (Hill Notation) C15H11I4NO4

  • Molecular Weight 776.87

  •  Beilstein/REAXYS Number 2954910

  •  MDL number MFCD00063056

  •  PubChem Substance ID 329826769

Purchase

Properties

assay   99%
mp   ~235 °C (dec.)
SMILES string   N[C@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(O)=O
InChI   1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m1/s1
InChI key   XUIIKFGFIJCVMT-GFCCVEGCSA-N
Gene Information   human ... THRA(7067), THRB(7068)
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

HPLC Analysis of Thyroxine Enantiomers on Astec® CHIROBIOTIC® TAG

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Thyroxine Enantiomers on Astec® CHIROBIOTIC® TAG
Keywords: Chromatography, Clinical, Forensic, High performance liquid chromatography, Hormones, Mass spectrometry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?