EMAIL THIS PAGE TO A FRIEND
V705 Sigma-Aldrich

L-Valine

99%

Synonym: (S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid

  • CAS Number 72-18-4

  • Linear Formula (CH3)2CHCH(NH2)CO2H

  • Molecular Weight 117.15

  •  Beilstein/REAXYS Number 1721136

  •  EC Number 200-773-6

  •  MDL number MFCD00064220

  •  PubChem Substance ID 329829061

Purchase

Properties

assay   99%
optical activity   [α]20/D +27.5°, c = 8 in 6 M HCl
optical purity   ee: 99% (GLC)
mp   295-300 °C (subl.) (lit.)
SMILES string   CC(C)[C@H](N)C(O)=O
InChI   1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1
InChI key   KZSNJWFQEVHDMF-BYPYZUCNSA-N

Description

Application

Reagent employed in organic synthesis including peptide coupling reactions.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YV9361000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

V0500 L-Valine, reagent grade, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?