• USA Home
  • W270700 - 6-Methyl-5-hepten-2-one

EMAIL THIS PAGE TO A FRIEND
W270700 Sigma-Aldrich

6-Methyl-5-hepten-2-one

natural, ≥98.0%, FCC

  • CAS Number 110-93-0

  • Linear Formula (CH3)2C=CHCH2CH2COCH3

  • Molecular Weight 126.20

  •  Beilstein/REAXYS Number 1741705

  •  EC Number 203-816-7

  •  MDL number MFCD00008905

  •  PubChem Substance ID 329830523

Purchase

Properties

Related Categories Alphabetical Listings, Certified Natural Products, Flavors and Fragrances, M-N
grade   natural
reg. compliance   FCC
assay   ≥98.0%
refractive index   n20/D 1.439 (lit.)
  n/D 1.440 (lit.)
bp   73 °C/18 mmHg (lit.)
density   0.858 g/mL at 20 °C
  0.848 g/mL at 25 °C (lit.)
  0.855 g/mL at 25 °C (lit.)
Organoleptic   citrus
SMILES string   C\C(C)=C\CCC(C)=O
InChI   1S/C8H14O/c1-7(2)5-4-6-8(3)9/h5H,4,6H2,1-3H3
InChI key   UHEPJGULSIKKTP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN1224 - class 3 - PG 3 - Ketones, liquid, n.o.s., HI: all
WGK Germany 
1
RTECS 
MJ9700000
Flash Point(F) 
132.8 °F
Flash Point(C) 
56 °C
Protocols & Articles

Articles

The Complex Regulatory Landscape for Natural Flavor Ingredients

Introduction The Regulatory Landscape US and EU Definitions of Natural Regional Regulatory Variations The Example of Vanillin The Case for Standardization
Mr. Michael Sabisch1, Mr. David Smith2

1Senior Quality Assurance Specialist
2International Sales Coordinator, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food Safety, Oxidations

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?