• USA Home
  • W305006 - Terpinyl isobutyrate, mixture of isomers

EMAIL THIS PAGE TO A FRIEND
W305006 Sigma-Aldrich

Terpinyl isobutyrate, mixture of isomers

≥85%

Synonym: p-menth-1-en-8-yl-isobutyrate

  • CAS Number 7774-65-4

  • Empirical Formula (Hill Notation) C14H24O2

  • Molecular Weight 224.34

  •  FEMA Number 3050

  •  EC Number 231-878-5

  •  Council of Europe no. 300

  •  MDL number MFCD00083088

  •  PubChem Substance ID 24901449

  •  Flavis number 9.425

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, Q-Z More...
biological source   synthetic
InChI Key   SMQUXKIIXFOJKI-UHFFFAOYSA-N
assay   ≥85%
refractive index   n20/D 1.461 (lit.)
bp   242 °C (lit.)
density   0.943 g/mL at 25 °C (lit.)
Organoleptic   fatty; pepper

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

New Product Offerings
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?