• USA Home
  • W305200 - Terpinyl formate, mixture of isomers

EMAIL THIS PAGE TO A FRIEND
W305200 Sigma-Aldrich

Terpinyl formate, mixture of isomers

≥97%, FG

Synonym: p-menth-1-en-8-yl-formate

  • CAS Number 2153-26-6

  • Empirical Formula (Hill Notation) C11H18O2

  • Molecular Weight 182.26

  •  FEMA Number 3052

  •  EC Number 218-444-0

  •  Council of Europe no. 348

  •  MDL number MFCD00036672

  •  PubChem Substance ID 24901450

  •  Flavis number 9.081

Purchase

Properties

Related Categories Alphabetical Listings, Certified Food Grade Products, Flavors and Fragrances, Halal Certified Products, Kosher Certified Products,
biological source   synthetic
grade   FG
  Halal
  Kosher
reg. compliance   EU Regulation 1334/2008 & 872/2012
assay   ≥97%
may contain   <0.10% alpha-tocopherol, synthetic as stabilizer
refractive index   n20/D 1.471 (lit.)
bp   220 °C (lit.)
density   0.981 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   floral; citrus; sweet
food allergen   no known allergens
SMILES string   CC1=CCC(CC1)C(C)(C)OC=O
InChI   1S/C11H18O2/c1-9-4-6-10(7-5-9)11(2,3)13-8-12/h4,8,10H,5-7H2,1-3H3
InChI key   IPYLQIQMGUZFCK-UHFFFAOYSA-N

Description

Packaging

1 kg in glass bottle

100 g in glass bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
OT0220000
Flash Point(F) 
206.6 °F
Flash Point(C) 
97 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?