• USA Home
  • W386006 - Ethyl methyl sulfide

EMAIL THIS PAGE TO A FRIEND
W386006 Sigma-Aldrich

Ethyl methyl sulfide

≥99%

  • CAS Number 624-89-5

  • Linear Formula CH3CH2SCH3

  • Molecular Weight 76.16

  •  FEMA Number 3860

  •  Beilstein/REAXYS Number 1696871

  •  EC Number 210-868-4

  •  MDL number MFCD00009268

  •  PubChem Substance ID 24901908

  •  Flavis number 12.154

Purchase

Properties

Related Categories Alphabetical Listings, E-F, Flavors and Fragrances More...
Quality Level   400
biological source   synthetic
vapor pressure   272 mmHg ( 37.7 °C)
assay   ≥99%
refractive index   n20/D 1.440 (lit.)
bp   66-67 °C (lit.)
mp   −106 °C (lit.)
density   0.842 g/mL at 25 °C (lit.)
Organoleptic   coffee; fishy; green; meaty; sulfurous
SMILES string   CCSC
InChI   1S/C3H8S/c1-3-4-2/h3H2,1-2H3
InChI key   WXEHBUMAEPOYKP-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Odor at 0.1%

Taste at 0.25-2 ppm

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
5 °F
Flash Point(C) 
-15 °C
Protocols & Articles

Articles

GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector, Size-exclusion chromatography

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

238317 Ethyl methyl sulfide, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?