EMAIL THIS PAGE TO A FRIEND
W502618 Sigma-Aldrich

DL-Alanine

FCC, ≥99%

Synonym: (±)-2-Aminopropionic acid

  • CAS Number 302-72-7

  • Linear Formula CH3CH(NH2)COOH

  • Molecular Weight 89.09

  •  Beilstein/REAXYS Number 1720250

  •  EC Number 206-126-4

  •  MDL number MFCD00064408

  •  PubChem Substance ID 329830735

  •  Flavis number 17.024

Purchase

Properties

biological source   synthetic
reg. compliance   FCC
assay   ≥99%
mp   289 °C (dec.) (lit.)
SMILES string   CC(N)C(O)=O
InChI   1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)
InChI key   QNAYBMKLOCPYGJ-UHFFFAOYSA-N

Description

Features and Benefits

Odorless, sweet taste

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1
Protocols & Articles

Articles

HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® T (No Mobile Phase Modifier)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® T (No Mobile Phase Modifier)
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® TAG

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® TAG
Keywords: Chromatography, High performance liquid chromatography

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?