• USA Home
  • W507504 - 4-Hydroxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND
W507504 Sigma-Aldrich

4-Hydroxybenzaldehyde

≥97%

  • CAS Number 123-08-0

  • Linear Formula HOC6H4CHO

  • Molecular Weight 122.12

  •  Beilstein/REAXYS Number 471352

  •  EC Number 204-599-1

  •  Council of Europe no. 0558

  •  MDL number MFCD00006939

  •  PubChem Substance ID 329830741

  •  Flavis number 5.047

Purchase

Properties

biological source   synthetic
assay   ≥97%
mp   112-116 °C (lit.)
Organoleptic   woody; sweet
SMILES string   [H]C(=O)c1ccc(O)cc1
InChI   1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H
InChI key   RGHHSNMVTDWUBI-UHFFFAOYSA-N

Description

Features and Benefits

Sweet, woody, balsamic

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
CU6475000
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?