• USA Home
  • Z500550 - Analtech TLC Uniplates: alumina matrix
EMAIL THIS PAGE TO A FRIEND
Z500550 Supelco
Analtech TLC Uniplates: alumina matrix
L × W 10 cm × 20 cm , scored
Purchase
Properties
Related Categories Analtech, Analtech Alumina Matrix, Analytical/Chromatography, Chromatography, Chromatography, TLC Plates,
material   alumina matrix
  glass support
feature   binder Inorganic
  fluorescent indicator
packaging   pkg of 25 ea
mfr. no.   Analtech, 04521
application(s)   thin layer chromatography (TLC): suitable
L × W   10 cm × 20 cm , scored
layer thickness   250 μm
Description
General description
Produced using a slurry containing calcium sulfate hemihydrate as a binder.
Other Notes
this product will be discontinued, please find the alternative product here :
105550
Legal Information
Uniplates is a trademark of Analtech, Inc.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?