• USA Home
  • Z566861 - IKA® RV 10 rotary evaporator, basic

EMAIL THIS PAGE TO A FRIEND
Z566861 Aldrich

IKA® RV 10 rotary evaporator, basic

vertical condenser, glass, AC/DC input 115 V AC, pack of 1 ea

Synonym: rotary evaporator condensers, rotovap condensers

Purchase

Properties

Related Categories Evaporators, IKA Rotary Evaporators, Labware More...
material   glass
feature   CSA approved
packaging   pack of 1 ea
parameter   20-280 rpm speed
W × H × D   500 mm × 430 mm × 410 mm
bath capacity   3 L
weight   19.6 kg

Description

General description

The IKA RV 10 rotary evaporators have integrated heating baths and an ergonomic, space-saving design. Each version features a motorized lifting mechanism that raises the evaporator piston out of the heating bath in case of power outage. Other safety features also include interval operation, smooth start, end position recognition for the evaporator piston, and a timer function.

The RV 10 evaporators are fitted with a water/oil bath that includes a powerful 1300 Watt heater capable of delivering temperatures from room temperature to 180°C. On the basic model, the temperature is controlled using a simple dial, and the basic system now comes with a drive capable of right-left interval operation as standard.

This produces significantly better results, especially in the case of drying processes. Highly solvent-resistant PTFE seals also ensure that operation of the system is exceptionally safe, even when used with aggressive organic solvents.

Basic model:
• The temperature is controlled using a simple dial
• Comes with a drive capable of right-left interval operation
• Solvent-resistant PTFE seals
• Supplied complete with a Woulff bottle to protect the vacuum pump against solvent condensation

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Protocols

Investigation of higher-order RNA G-quadruplex structures in vitro and in living cells by 19F NMR spectroscopy

a Division of Chemistry, Department of Medical Sciences, Faculty of Medicine, University of Miyazaki
Keywords: Cancer, Capillary electrophoresis, Cell disruption, Chromatography, Click chemistry, Electron spin resonance, Gene expression, High performance liquid chromatography, Indicators, Industries, Ligands, Liquid chromatography mass spectrometry, Mass spectrometry, Melting, Methylations, Nuclear magnetic resonance spectroscopy, Nucleic acid annealing, Protein assay, Purification, RNA purification, Separation, Solvents, Spectroscopy, Thin layer chromatography, Transcription

Related Content

IKA Evaporators [VIDEO]

From our library of Related Content, Sigma-Aldrich presents IKA Evaporators [VIDEO]

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?