• USA Home
  • Z639036 - Bollé Targa safety eyeshields

EMAIL THIS PAGE TO A FRIEND
Z639036

Bollé Targa safety eyeshields

yellow frame, clear polycarbonate lens

  •  NACRES NB.35

Purchase

Properties

material   clear polycarbonate lens
grade   CE Compliant
mfr. no.   Bollé® TAPSI
availability   not available in USA and Canada
color   yellow frame

Description

General description

• Pivoting temples
• Anti-scratch & anti-fog

Legal Information

Bollé is a registered trademark of Bushnell Performance Optics Europe Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?