• USA Home
  • Z679801 - Chemrus® disposable filter funnel
EMAIL THIS PAGE TO A FRIEND
Z679801
Chemrus® disposable filter funnel
Buchner, pore size 10 μm, capacity 40-60 mL
  •  NACRES NB.43
Purchase
Properties
material   PE frit
  polypropylene body
mfr. no.   Chemrus, CR-1018-40
capacity   40-60 mL
pore size   10 μm
Description
General description
Convenient and inexpensive, these disposable filter funnels have polypropylene bodies with tapered stems that fit 7/10 joints without seals. The funnels are available in three styles, Buchner and Hirsch, and conical, choice of polyethylene frit, perforated plate, acid washed Celite or silica gel. Maximum use temperature is 110 °C. Funnels have an 80 mm long stem so the glass vacuum adapters are not contaminated during the filtration process.
Order vacuum adapters, filter paper, vials and 20-400 to 24-400 vial connectors separately.
Disposable filter funnels with a polypropylene body with a 10 micron polyethylene fritted disc (40 micron disc on the 500 mL funnel) or choice of acid washed Celite or silica gel.
Legal Information
Chemrus is a registered trademark of Liu, Xioagao & Cai, Jianjian
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?