• USA Home
  • ERS-020 - Internal Standards Mixture

EMAIL THIS PAGE TO A FRIEND
ERS-020 Supelco

Internal Standards Mixture

4000 μg/mL each component in methylene chloride, ampule of 1.2 mL, certified reference material, Cerilliant®

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Certified Reference Materials from Cerilliant,
grade   certified reference material
feature   SNAP-N-SPIKE®/SNAP-N-SHOOT®
composition   Acenaphthene-D10
  Chrysene-D12
  1,4-Dichlorobenzene-D4
  Naphthalene-d8
  Perylene-D12
  Phenanthrene-D10
packaging   ampule of 1.2 mL
mfr. no.   Cerilliant®
concentration   4000 μg/mL each component in methylene chloride
Featured Industry   Environmental
format   multi-component solution
storage temp.   room temp

Description

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Snap-N-Shoot is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Analyte

Description

Product #

Add to Cart

1,4-Dichlorobenzene-d4    
Acenaphthene-d10    
Chrysene-d12    
Naphthalene-d8    
Perylene-d12    
Phenanthrene-d10    
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

CRM47906 EPA 8270 Semivolatile Internal Standard Mix, High Concentration, certified reference material, 4000 μg/mL each component in methylene chloride: benzene (1:1), ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?