EMAIL THIS PAGE TO A FRIEND
L-042 Supelco

Lithocholic acid

50 μg/mL in methanol, certified reference material, Cerilliant®

Synonym: 3α-Hydroxy-5β-cholan-24-oic acid, 3α-Hydroxy-5β-cholanic acid, 5β-Cholan-24-oic acid-3α-ol

  • CAS Number 434-13-9

  • Empirical Formula (Hill Notation) C24H40O3

  • Molecular Weight 376.57

  •  Beilstein/REAXYS Number 3217757

  •  EC Number 200-659-6

  •  MDL number MFCD00003682

  •  PubChem Substance ID 329817364

  •  NACRES NA.24

Purchase

Properties

Quality Level   300
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   50 μg/mL in methanol
mp   183-188 °C (lit.)
format   single component solution
storage temp.   −20°C
SMILES string   [H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC(O)=O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@@H](O)C2
InChI   1S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25H,4-14H2,1-3H3,(H,26,27)/t15-,16-,17-,18+,19-,20+,21+,23+,24-/m1/s1
InChI key   SMEROWZSTRWXGI-HVATVPOCSA-N

Description

General description

This Certified Spiking Solution® is suitable for use as starting material in the preparation of linearity standards, calibrators, and controls in LC-MS/MS and GC/MS bile acid testing methods. Lithocholic acid (LCA) is a bile acid that acts as a detergent to solubilize fats for absorption in the intestine. Mass spectrometry-based analysis of LCA is routinely performed in clinical diagnostic testing applications including neonatal testing of inborn errors of bile acid synthesis and differentiating among types of familial intrahepatic cholestasis.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation

Snap-N-Shoot is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Eyes
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
WGK 2
Flash Point(F) 
49.5 °F - closed cup
Flash Point(C) 
9.7 °C - closed cup
Protocols & Articles

Articles

HPLC Analysis of Bile Acids on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Bile Acids on Ascentis® Express C18
Keywords: Chromatography, Clinical, High performance liquid chromatography

TLC analysis of Cholesterol and its Bile Acid Metabolites on HPTLC pre-coated plate RP-2 F 254s

From our library of Articles, Sigma-Aldrich presents TLC analysis of Cholesterol and its Bile Acid Metabolites on HPTLC pre-coated plate RP-2 F 254s

Related Products

related product

Product #

Description

Add to Cart

C-146 Cholic acid, 500 μg/mL in methanol, certified reference material, Cerilliant®
U-002 Ursodeoxycholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®
C-147 Chenodeoxycholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?