• USA Home
  • T-098 - Testosterone Calibrator

EMAIL THIS PAGE TO A FRIEND
T-098 Supelco

Testosterone Calibrator

Level 3, 9 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant®

Synonym: Testosterone Calibrator

  • Empirical Formula (Hill Notation) C19H28O2

  • Molecular Weight 288.42

  •  NACRES NA.24

Purchase

Properties

Quality Level   300
grade   certified reference material
packaging   pkg of 1 mL
mfr. no.   Cerilliant®
IVD   for in vitro diagnostic use
concentration   9 (ng/dL, Stripped Serum)
format   single component solution
shipped in   dry ice
storage temp.   −20°C

Description

General description

Certified reference material of testosterone in stripped serum, CE marked for in vitro diagnostic (IVD) applications by mass spectrometry. Individual levels of testosterone in stripped serum are offered separately at concentrations ranging from 2 to 2,000 ng/dL. Each individual level is ready to use as a calibrator in LC-MS/MS testosterone methods. Separate options for a blank and 13C- or deuterium labeled internal standards of testosterone or the whole kit including calibrators, blank and Testosterone-2,3,4-13C3 in 10 μg/mL Acetonitrile are also available.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Testosterone Serum Calibrator Kit - CE Mark

Are you in the United States or Canada ? Please view our page on Certified Reference Material of Testosterone in Serum
Keywords: Cancer, Capillary electrophoresis, Clinical, Diagnostic, High performance liquid chromatography, Hormones, Ion-exchange chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, PAGE

Related Products

related product

Product #

Description

Add to Cart

T-095 Testosterone-2,3,4-13C3 solution, 10 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
D-077 5α-Dihydrotestosterone-D3 (16,16,17-D3) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-058 Epitestosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
H-059 6β-Hydroxytestosterone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
M-906 17α-Methyltestosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®
T-034 6β-Hydroxytestosterone-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
T-037 Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-046 Testosterone-d3 (16, 16, 17-d3) solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-070 Testosterone-2,3,4-13C3 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?