• USA Home
  • T-107 - Testosterone Calibrator

EMAIL THIS PAGE TO A FRIEND
T-107 Supelco

Testosterone Calibrator

12 mL (12 x1 mL; Kit contains 1 ampoule (1 mL) each of T-095, Testosterone Calibrator Level 1 through 10, Testosterone Calibrator Blank), pkg of 12 mL, Cerilliant®

Synonym: Testosterone Calibrator

  • Empirical Formula (Hill Notation) C19H28O2

  • Molecular Weight 288.42

  •  NACRES NA.24

Purchase

Properties

Quality Level   300
grade   certified reference material
packaging   pkg of 12 mL
mfr. no.   Cerilliant®
IVD   for in vitro diagnostic use
concentration   12 mL (12 x1 mL; Kit contains 1 ampoule (1 mL) each of T-095, Testosterone Calibrator Level 1 through 10, Testosterone Calibrator Blank)
format   multi-component solution
shipped in   dry ice
storage temp.   −20°C

Description

General description

The Testosterone Calibrator Kit includes certified reference material of testosterone at concentrations ranging from 2 to 2,000 ng/dL in stripped serum, stripped serum blank and testosterone-2,3,4-13C3 in 10 μg/mL acetonitrile as internal standard. The kit is CE marked and suitable for in vitro diagnostic (IVD) applications by mass spectrometry. Individual levels of testosterone in stripped serum, serum blank and internal standard are also available separately.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Kit component also available separately

Description

Product #

Add to Cart

Testosterone Calibrator, Level 1, 2 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® SDS T-096
Testosterone Calibrator, Level 2, 4 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® 1mL SDS T-097
Testosterone Calibrator, Level 3, 9 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® 1 mL SDS T-098
Testosterone Calibrator, Level 4, 17.5 ( ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® SDS T-099
Testosterone Calibrator, Level 5, 35 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® SDS T-100
Testosterone Calibrator, Level 6, 52.5 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® SDS T-101
Testosterone Calibrator, Level 7, 150 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® SDS T-102
Testosterone Calibrator, Level 8, 500 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® SDS T-103
Testosterone Calibrator, Level 9, 750 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® SDS T-104
Testosterone Calibrator, Level 10, 2000 ( ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant® SDS T-105
Safety & Documentation

Safety Information

RIDADR 
UN 1648 3 / PGII

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Testosterone Serum Calibrator Kit - CE Mark

Are you in the United States or Canada ? Please view our page on Certified Reference Material of Testosterone in Serum
Keywords: Cancer, Capillary electrophoresis, Clinical, Diagnostic, High performance liquid chromatography, Hormones, Ion-exchange chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, PAGE

Related Products

related product

Product #

Description

Add to Cart

T-095 Testosterone-2,3,4-13C3 solution, 10 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
D-073 5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
D-077 5α-Dihydrotestosterone-D3 (16,16,17-D3) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-058 Epitestosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
H-059 6β-Hydroxytestosterone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
M-906 17α-Methyltestosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®
T-034 6β-Hydroxytestosterone-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
T-037 Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-046 Testosterone-d3 (16, 16, 17-d3) solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-070 Testosterone-2,3,4-13C3 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?