EMAIL THIS PAGE TO A FRIEND
A-132 Cerilliant

Acyclovir solution

1.0 mg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material

  • Empirical Formula (Hill Notation) C8H11N5O3

  • Molecular Weight 225.20

Purchase

Properties

grade   certified reference material
packaging   ampule of 1 mL
concentration   1.0 mg/mL in methanol: water (1:1)
format   single component solution
storage temp.   2-8°C
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

E-109 Efavirenz solution, 1.0 mg/mL in acetonitrile, certified reference material
N-098 Nevirapine solution, 1.0 mg/mL in methanol, certified reference material
R-022 Ritonavir solution, 1.0 mg/mL in acetonitrile, certified reference material
Z-011 Zidovudine solution, 1.0 mg/mL in methanol, certified reference material
G-020 Ganciclovir, 1.0 mg/mL (1% 1M HCl in Methanol), certified reference material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?