• USA Home
  • H-115 - 25H-NBOMe hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
H-115 Cerilliant

25H-NBOMe hydrochloride solution

1.0 mg/mL in methanol, certified reference material, ampule of 1 mL

  • CAS Number 1566571-52-5

  • Empirical Formula (Hill Notation) C18H23NO3 · HCl

  • Molecular Weight 337.84

Purchase

Properties

grade   certified reference material
InChI Key   OOFBPLRIHRHTBL-UHFFFAOYSA-N
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
concentration   1.0 mg/mL in methanol
format   single component solution
storage temp.   −20°C

Description

General description

25H-NBOMe is a second generation 2C amine derivative of the phenethylamine 2C-H. Sold under street names such as New Nexus, N-Bomb, and 2CH this designer drug is a potent hallucinogen with active doses in the microgram range. Suitable uses for this certified Snap-N-Spike® solution include as starting material for calibrators or controls in 25H-NBOMe testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.

Legal Information

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
2
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

B-061 25B-NBOMe hydrochloride solution, 1.0 mg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material
B-062 25B-NBOMe-D3 hydrochloride solution, 100 μg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material
I-016 25I-NBOMe hydrochloride solution, 1.0 mg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material
I-017 25I-NBOMe-D3 hydrochloride solution, 100 μg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material
C-131 25C-NBOMe hydrochloride solution, 1.0 mg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material
C-132 25C-NBOMe-D3 hydrochloride solution, 100 μg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material
I-019 25I-NBOH hydrochloride solution, 1.0 mg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material
I-020 25I-NBF hydrochloride solution, 1.0 mg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material
I-022 25I-NBMD hydrochloride solution, 1.0 mg/mL in methanol ((as free base)), ampule of 1 mL, certified reference material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?