• USA Home
  • SCGPT10RE - Steritop Threaded Bottle Top Filter

EMAIL THIS PAGE TO A FRIEND
SCGPT10RE Millipore

Steritop Threaded Bottle Top Filter

Sterile, 1000 mL process volume, 0.22 µm pore size, GP, 45 mm, polyethersulfone membrane, pkg of 12

  •  eCl@ss 32031623

  •  NACRES NB.22

Purchase

Properties

packaging   12 pack, GP, 0.22 µm, polyethersulfone, 1000 mL 45 mm, radio-sterilized (SCGPT10RE)
H   169 mm
application(s)   sterile filtration: suitable
diam.   13.5 cm
feature   holdup volume 2.4 mL
filter diam.   45 mm
filtration area   40 cm2
fitting   inlet funnel
  outlet thread(s) 45 mm
funnel capacity   1 L
material   polyethersulfone membrane
  polystyrene
matrix   Express PLUS
parameter   1000 mL process volume
  45 °C max. temp.
pore size   0.22 μm nominal pore size
  0.22 μm pore size
shipped in   ambient
test parameters   sample volume: 1 L
trade name   Steritop
sterility   sterile
wettability   Hydrophilic

Description

Application

Sterile, 1000 mL process volume, 0.22 µm pore size, GP, 45 mm, polyethersulfone membrane, pkg of 12

General description

The Stericup® vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format. Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device. The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications

Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component

Principle

Vacuum

Preparation Note

Sterilization Method
Gamma irradiation

Physical form

Color Code: Clear with blue adapter

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Stem Cell Testing of PES Membrane Containing Stericup® Quick Release Filters

Though microporous filtration is the method of choice to prepare and minimize contamination of cell culture media and reagents, there still remains some concern that the filtration step will remove v...
Keywords: Buffers, Capture ELISA, Cell culture, Culture media, Enzyme-linked immunosorbent assay, Filtration, Gene expression, Growth factors, Immunocytochemistry, Sterilizations, Transcription

Related Content

Filter Membrane Selection Recommendations

What are you filtering out? What do you want to keep? Pick the right membrane for your filtration.
Keywords: Cell culture, Filtration

Stericup® and Steritop® Filtration Systems

Stericup® Sterile Vacuum Filtration Systems are known to research scientists worldwide for reliable, high-performance membranes and optimum sterile filtration of laboratory reagents, buffers, and med...
Keywords: Buffers, Culture media, Filtration, Sterilizations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?