• USA Home
  • LUMFILM-RO - Lumi-Film Chemiluminescent Detection Film

EMAIL THIS PAGE TO A FRIEND
LUMFILM-RO Roche

Lumi-Film Chemiluminescent Detection Film

8 x 10 inches, 20.3 x 25.4 cm

Synonym: detection film, lumi-film | lumi-film chemiluminescent detection film

Purchase

Properties

Related Categories Chemiluminescent, Molecular Biology, Nucleic Acid Detection Substrates and Reagents, Nucleic Acid Detection and Hybridization
Quality Level   100
packaging   pkg of 100 (11666657001)
  pkg of 100 (11666916001)
mfr. no.   Roche
film L × W   7.1 in. (18 cm) × 9.4 in. (24 cm), 11666916001
  8 in. (20.3 cm) × 10 in. (25.4 cm), 11666657001
shipped in   ambient
storage temp.   20-25°C
  20-25°C

Description

General description

The film is not interleaved.

Film is all in one box.

Application

Lumi-Film Chemiluminescent Detection Film is ideal for detecting the signals from alkaline phosphatase chemiluminescent substrates (e.g., CSPD) in membrane hybridization techniques.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Lumi-Film Chemiluminescent Detection Film Troubleshooting

Possible causes: Inappropriate membrane handling during procedure. Note: Every scratch, touch, or gouge on the surface of the membrane will be made visible during the chemiluminescent detection/visua...
Keywords: Nucleic acid hybridization

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?