EMAIL THIS PAGE TO A FRIEND
00190180 Sigma-Aldrich

Hypericin

primary reference standard

Synonym: 4,5,7,4′,5′,7′-Hexahydroxy-2,2′-dimethylnaphthodianthrone

  • CAS Number 548-04-9

  • Empirical Formula (Hill Notation) C30H16O8

  • Molecular Weight 504.44

  •  Beilstein Registry Number 1917913

  •  EC Number 208-941-0

  •  MDL number MFCD00016683

Purchase

Properties

grade   primary reference standard
InChI Key   BTXNYTINYBABQR-UHFFFAOYSA-N
CofA   certificate is enclosed in each package
shelf life   limited shelf life, expiry date on the label
storage temp.   −20°C

Description

Application

Reference standard in the analysis of herbal medicinal products.

General description

Produced by HWI Analytik
Exact content can be found on the certificate

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?