EMAIL THIS PAGE TO A FRIEND
00303 Sigma-Aldrich

Deoxynivalenol

≥97.0% (TLC)

Synonym: 3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin

  • CAS Number 51481-10-8

  • Empirical Formula (Hill Notation) C15H20O6

  • Molecular Weight 296.32

  •  MDL number MFCD09039270

Purchase

Properties

InChI Key   LINOMUASTDIRTM-QGRHZQQGSA-N
assay   ≥97.0% (TLC)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
YD0167000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D0156 Deoxynivalenol

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?