EMAIL THIS PAGE TO A FRIEND
00486 Sigma-Aldrich

Carbon dioxide

puriss., ≥99.998%

  • CAS Number 124-38-9

  • Empirical Formula (Hill Notation) CO2

  • Molecular Weight 44.01

  •  Beilstein/REAXYS Number 1900390

  •  EC Number 204-696-9

  •  MDL number MFCD00011491

  •  PubChem Substance ID 329747082

Purchase

Properties

Related Categories Chemical Synthesis, Specialty Gases, Synthetic Reagents More...
vapor density   1.52 (vs air)
vapor pressure   56.5 atm ( 20 °C)
InChI Key   CURLTUGMZLYLDI-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.998%
mp   −78.5 °C (lit.)

Description

Packaging

pressure tin
filling pressure at 15°C: 11 bar; content (15°C, 1 bar): 11 l

Other Notes

Sales restrictions may apply

Safety & Documentation

Safety Information

RIDADR 
UN 1013 2.2
WGK Germany 
-
RTECS 
FF6400000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

295108 Carbon dioxide, ≥99.8%

control valve

Product #

Description

Add to Cart

97490 Flow controlling valve for 600 mL and 1000 mL pressure tins, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?