• USA Home
  • 00888 - 9-Anthracenecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
00888

9-Anthracenecarboxylic acid

puriss. p.a., matrix substance for MALDI-MS, ≥99.0% (HPLC)

Synonym: 9-Anthroic acid

  • CAS Number 723-62-6

  • Empirical Formula (Hill Notation) C15H10O2

  • Molecular Weight 222.24

  •  Beilstein/REAXYS Number 1875336

  •  EC Number 211-964-9

  •  MDL number MFCD00001257

  •  PubChem Substance ID 329747157

Purchase

Properties

grade   matrix substance for MALDI-MS
  puriss. p.a.
assay   ≥99.0% (HPLC)
impurities   ≤0.5% ash
mp   213-217 °C (lit.)
solubility   chloroform/methanol (9:1): 10 mg/mL, clear
cation traces   Ba: ≤5 mg/kg
  Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤500 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg
fluorescence   λex 255 nm; λem 411 nm in ethanol
SMILES string   OC(=O)c1c2ccccc2cc3ccccc13
InChI   1S/C15H10O2/c16-15(17)14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H,(H,16,17)
InChI key   XGWFJBFNAQHLEF-UHFFFAOYSA-N

Description

Analysis Note

In addition to the emission maximum at 411 nm, there are lower maxima at 388, 435 and 458 nm.

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A89405 9-Anthracenecarboxylic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?