EMAIL THIS PAGE TO A FRIEND
01670 Sigma-Aldrich

Acridone

purum, for fluorescence, ≥98.0% (HPLC)

Synonym: 9,10-Dihydro-9-oxoacridine

  • CAS Number 578-95-0

  • Empirical Formula (Hill Notation) C13H9NO

  • Molecular Weight 195.22

  •  Beilstein Registry Number 7104

  •  EC Number 209-434-7

  •  MDL number MFCD00005019

  •  PubChem Substance ID 24844904

Purchase

Properties

grade   for fluorescence
InChI Key   FZEYVTFCMJSGMP-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   >300 °C (lit.)
Gene Information   human ... ABCB1(5243)
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
AR6976000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?