EMAIL THIS PAGE TO A FRIEND
02250 Sigma-Aldrich

(−)-Epinephrine

≥97.0% (NT)

Synonym: (−)-Adrenalin, (R)-(−)-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol, L-Adrenaline, L-Epinephrine

  • CAS Number 51-43-4

  • Linear Formula (HO)2C6H3CH(OH)CH2NHCH3

  • Molecular Weight 183.20

  •  Beilstein/REAXYS Number 2368277

  •  EC Number 200-098-7

  •  MDL number MFCD00002204

  •  PubChem Substance ID 329747406

Purchase

Properties

assay   ≥97.0% (NT)
optical activity   [α]20/D −52±2°, c = 5% in 5% HCl
ign. residue   ≤0.1%
loss   ≤1% loss on drying
mp   215 °C (dec.) (lit.)
  ~215 °C (dec.)
storage temp.   2-8°C
SMILES string   CNC[C@H](O)c1ccc(O)c(O)c1
InChI   1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3/t9-/m0/s1
InChI key   UCTWMZQNUQWSLP-VIFPVBQESA-N

Description

Biochem/physiol Actions

Adrenoceptor agonist.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
DO2625000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?